Chemical-Resistant PP Labels B-7425J for BradyJet™ J7300 Inkjet Printer
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
176606 176635 176584 176613 176611 176585 176609 176633 176615 176607 176631 170479 176634 170463 176610 176614 176617 176616 170464 176618 176608 176636 170478
BRDY176606EA 133.28 EUR
BRDY176606 BRDY176635 BRDY176584 BRDY176613 BRDY176611 BRDY176585 BRDY176609 BRDY176633 BRDY176615 BRDY176607 BRDY176631 BRDY170479 BRDY176634 BRDY170463 BRDY176610 BRDY176614 BRDY176617 BRDY176616 BRDY170464 BRDY176618 BRDY176608 BRDY176636 BRDY170478
Chemical-Resistant PP Labels B-7425J for BradyJet™ J7300 Inkjet Printer
Etiketten
Create full-colour identification and barcode labels B-7425J that outlast liquid nitrogen, autoclave conditions, and harsh chemicals. For use on smooth surfaces.
  • Labels withstand extreme temperatures from −80 to +70 °C
  • Permanent acrylic adhesive bonds tightly to smooth surfaces like glass, copper, and stainless steel
  • Inkjet receptive polypropylene material
  • Designed for indoor use
  • Abrasion-resistant, low temperature resistance, solvent/chemical resistant
  • Matt finish
Inkjet receptive label material is ideal for full-colour laboratory identification onMore Product Information