Axygen® HybridRack Pipette Tips
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ERF-10XT-R-S ERF-200NX-L-RS ERF-200NX-R-S ERF-20-L-R-S ERF-20-R-S ER-1000XT-R-S ER-10XT-R-S ER-200-R-S ER-300NX-R-S ER-300XT-R-S ERF-1000XT-L-R ERF-1000XT-R-S ERF-100-L-R-S ERF-100-R-S ERF-10XT-L-R-S ERF-300XT-L-RS ERF-300XT-R-S
613-7395EA 113 EUR
613-7395 613-7407 613-7398 613-7393 613-7396 613-7405 613-7401 613-7402 613-7403 613-7404 613-7406 613-7400 613-7394 613-7397 613-7392 613-7408 613-7399
Axygen® HybridRack Pipette Tips
Pipettips
Filtered, maxymum recovery, and sterile pipette tips in a truly sustainable packaging format that reduces plastic by 70% per case.
  • Patent pending design uses 70% less plastic than traditional tip rack, equal to six pounds of plastic saved per case
  • Paperboard base is easy to recycle with most local recycling programs
  • Made in the USA in an EngergyStar, ISO 14001, and ISO 13485 certified facility
  • Combination of the strong plastic deck and sturdy paperboard base allow for pipetting directly from the HybridRack, even with a mulit-channel instrument
  • Colour coded decks depending on tip volume
The HybridRack packaging format exchanges the plastic base of the pipette tip rack with a high quality, fully recyclable, paperboard base. It utilises the polypropylene deck from our established and trusted Axygen MultiRack and Corning DeckWorks product lines in conjunction with the new paperboard base to create a stable, sustainable pipetting platform. End users can pipet directly from the HybridRack with a single or multi-channel pipettor, finish a rack of tips and simply recycle the HybridRack in their curbside recycling bin.
Using HybridRack, significantly less waste is generated from pipette tips racks: Compared to traditional plastic pipette tip racks, each case of 200 µl tips in the HybridRack packaging format saves six pounds of plastic. For 1000 µl tips, eight pounds of plastic is saved! With HybridRack, Corning takes the effort out of being sustainable so customers can focus on what really matters: their research.
Certificaten: ISO 13485, ISO 14001, EnergyStar.
Verpakking: Packaged in Axygen's newly redesigned sustainable pack box that uses less ink, printed on 100% recycled corrugate with an aqueous coating.