Membrane Filters, Nylon
Leverancier: Advantech
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N500A047A N500A013A
ADVTN500A047AEA 339 EUR
ADVTN500A047A ADVTN500A013A
Membrane Filters, Nylon
Filters Membraanfilters
These membrane filters are inherently hydrophilic membranes made by impregnating a polyester web with nylon and are used for HPLC sample preparation, to sterilize biological solutions or buffers, or for vacuum degassing.
  • High strength and heat resistance
  • Binds proteins, DNA and RNA
  • Pure, negligible organic extractables
  • Maximum operating temperature: 180 °C
  • Compatible with many aqueous and alcoholic solvents and solutions
Suitable for HPLC sample preparation. Filter sterilise and clarify aqueous and organic solvent solutions including buffers, microbiological and tissue culture solutions. Vacuum degassing.