Cole-Parmer® Luer to Hose Barb Fittings
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
50110-11 50110-21
COLE50110-11EA 0 EUR
COLE50110-11 COLE50110-21
Cole-Parmer® Luer to Hose Barb Fittings
Fittingen en connectoren
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure the integrity is not compromised. An ISO Class 7 cleanroom utilises HEPA filtration systems to maintain air cleanliness levels of less than 10000 particles of 0,5 µm.
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine
More Product Information