epT.I.P.S.® Eppendorf Quality™ BioBased Standard Pipette Tips, Bulk
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0030080.033 0030080.041 0030080.050 0030080.068 0030080.076 0030080.084 0030080.092 0030080.106 0030080.114 0030080.122 0030080.130
EPPE0030080.033EA 72.5 EUR
EPPE0030080.033 EPPE0030080.041 EPPE0030080.050 EPPE0030080.068 EPPE0030080.076 EPPE0030080.084 EPPE0030080.092 EPPE0030080.106 EPPE0030080.114 EPPE0030080.122 EPPE0030080.130
epT.I.P.S.® Eppendorf Quality™ BioBased Standard Pipette Tips, Bulk
Pipettips
Improve your laboratory carbon footprint and switch to epT.I.P.S.® BioBased. These pipette tips are produced from recycled feedstock in second generation, derived solely from waste and residue streams from vegetable oil production. epT.I.P.S.®.
  • Original, high-quality Eppendorf pipette tips packaged in resealable bags
  • 100% Bio-based polypropylene: Produced from renewable resources
  • Uncompromised Performance: Identical performance properties and premium quality as our fossil-based epT.I.P.S.®
  • ISCC Certified: Made from plastic that can be traced back to biological waste materials
  • Premium material, quality and purity certified
  • Get excellent sample integrity as no slip agents, plasticizers and biocides are used
  • Independent laboratory – Lot-specific purity certificates can be downloaded from Eppendorf website
BioBased Bulk packs offer a practical, high-volume solution in 'Eppendorf Quality', ensuring precise and reliable measurements.