Nitro blue tetrazolium chloride (NBT) tech. 90%, Technical Grade
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride
B23792.02 B23792.88
B23792.02EA 83.1 EUR
B23792.02 B23792.88
Nitro blue tetrazolium chloride (NBT) tech. 90%, Technical Grade
Nitro blue tetrazolium chloride (NBT)
Purity: 90% CAS No.: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Formula Weight: 817.65 Melting Point: ca 190° dec. Beilstein Reference: 4115923
Formule: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Smeltpunt: ∼ 190 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012159
CAS nummer: 298-83-9
EINECS: 206-067-4