Nitro blue tetrazolium chloride (NBT), Ultrapure
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride
0329-1GEA 193 EUR
0329-1G 0329-500MG
Nitro blue tetrazolium chloride (NBT), Ultrapure
Nitro blue tetrazolium chloride (NBT)
Formule: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Smeltpunt: 200
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00012159
CAS nummer: 298-83-9
EINECS: 206-067-4
Order Now
Specification Test Results
Em (Lambda Max, Methanol) >54000
Lambda Max (Methanol) 255 – 260 nm
Loss on Drying <10.0%
Solubility (1%, Water) Pass