Chromotropic acid disodium salt dihydrate ≥98.5% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate, Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate

D0596-25G D0596-500G
TCIAD0596-25GEA 53.9 EUR
TCIAD0596-25G TCIAD0596-500G
Chromotropic acid disodium salt dihydrate ≥98.5% (by titrimetric analysis)
Chromotropic acid disodium salt dihydrate
Formule: (HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MW: 400.29 g/mol
Smeltpunt: 300
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150612
CAS nummer: 5808-22-0
EINECS: 204-972-9
Merck Index: 13,02261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f9c9d1eafb06879f81097982f90fb840d6fd9a0&vid=9f9c9d1eafb06879f81097982f90fb840d6fd9a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystaline
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.5 % (calcd.on anh.substance)
Water 8.0 to 11.0 %
Sulfate max. 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR