AlphaTec® 87-224 Industrial Gloves
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87-224/7 87-224/8 87-224/9 87-224/10
ANSE87-224/7EA 44.5 EUR
ANSE87-224/7 ANSE87-224/8 ANSE87-224/9 ANSE87-224/10
AlphaTec® 87-224 Industrial Gloves
Handschoenen
Two-tone double-dipped work gloves, with a combination of neoprene and natural rubber.
Specialised protection: AlphaTec® 87-224 industrial gloves are two-toned and double-dipped, blending neoprene and natura rubber to offer personal protection against various chemicals
Enhanced defenses: Thanks to their mediumweight thickness of 0,68 mm/27 mil, they also provide a robust barrier against mechanical hazards
Elevated comfort: Designed with 100% cotton flock lining, these sturdy work gloves absorb wearers’ sweat, for greater comfort with sustained use
Strengthened grip: These AlphaTec® safety gloves also feature a honeycomb pattern finish, boosting grip on wet and dry surfaces
Recommended for: Maintenance of plant and heavy equipment, laboratory maintenance and clean up, janitorial use, chemical handling
Primary industries: Automotive aftermarket, machinery and equipment, infrastructure maintenance, janitorial/sanitation, chemical, utilities, warehousing