Dicyclohexylcarbodiimide ≥99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1,3-Dicyclohexylcarbodiimide, DCC, N,N'-Dicyclohexylcarbodiimide
A10973.14 A10973.22 A10973.36
A10973.14EA 23.6 EUR
A10973.14 A10973.22 A10973.36
Dicyclohexylcarbodiimide ≥99%
Dicyclohexylcarbodiimide
Purity: 99% CAS No.: 538-75-0 Molecular Formula: C13H22N2 Formula Weight: 206.33 Flash Point: 138°(280°F) Melting Point: 33-36° Boiling Point: 154-156°/11mm Beilstein Reference: 610662 Merck Reference: 14,3096 UN No.: 2811 MDL No.: MFCD00011659
Formule: C₁₃H₂₂N₂
MW: 206.33 g/mol
Kookpunt: 154…156 °C (11 mmHg)
Smeltpunt: 33…36 °C
Dichtheid: 0.95 g/cm³ (40 °C)
Vlampunt: 138 °C (280 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011659
CAS nummer: 538-75-0
EINECS: 208-704-1
UN: 2811
ADR: 6.1 III
Merck Index: 13,03123