Florisil®, powder_fine <200 mesh Chromatography, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Florisil
MQ100
288705-250G 288705-1KG
288705-250GEA 112 EUR
288705-250G 288705-1KG
Florisil®, powder_fine <200 mesh Chromatography, Supelco®
Magnesium silicate
Formule: MgO₃Si
MW: 100.38 g/mol
MDL Nummer: MFCD00078249
CAS nummer: 1343-88-0
EINECS: 215-681-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7185f1253a0288e3ff39ccac6243db3984fe6de6&vid=7ba8b51c22ac5608f10c5b1ec5b0df6942a86b2a&prid=web&clid=SAPDAM