Florisil® Chromatography, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Florisil
MQ200
46385-500G-F 46385-5KG-F
46385-500G-FEA 261 EUR
46385-500G-F 46385-5KG-F
Florisil® Chromatography, Supelco®
Magnesium silicate
Formule: MgO₃Si
MW: 100.38 g/mol
MDL Nummer: MFCD00078249
CAS nummer: 1343-88-0
EINECS: 215-681-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2c888e653111b8d5ed2a2a4529f3776d5914dc6&vid=6afdd8244a091087ffbdc0eb4fc12bf74ec30a92&prid=web&clid=SAPDAM