N-Octyl-β-D-glucopyranoside
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside
31055.02
31055.03
SERA31055.02EA
0
EUR
SERA31055.02
SERA31055.03
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
Formule:
C₁₄H₂₈O₆ MW: 292.37 g/mol Smeltpunt: 110 Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00063288 CAS nummer: 29836-26-8 EINECS: 249-887-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...