VWR® AVS TITRINORM, ntimony Standard Solution, 1000 mg/L Sb in 5% Nitric Acid with Hydrofluoric Acid (max. 1%), Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86707.260EA 23.31 EUR
86707.260 86707.180
VWR® AVS TITRINORM, ntimony Standard Solution, 1000 mg/L Sb in 5% Nitric Acid with Hydrofluoric Acid (max. 1%), Standard for AAS
Standaarden Kationische standaarden Antimonium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%