VWR®, Arsenic standard solution, 1,000 mg/L As in dil. nitric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86661.260EA 36.1 EUR
86661.260 86661.180
VWR®, Arsenic standard solution, 1,000 mg/L As in dil. nitric acid AVS TITRINORM standard for AAS
Standaarden Kationische standaarden Arseen standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%