Molykote® High vacuum grease
Leverancier: DOW CORNING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
636082B
291-0039EA 42.2 EUR
291-0039
Molykote® High vacuum grease
Smeermiddelen
Light grey, translucent, non melting silicone. Serviceable from –40 to +260 °C.
Leveringsinformatie: This product is not available in all countries. Please check with your local Avantor sales office.