VWR®, Manganese standard solution, 1000 mg/L Mn in dil. nitric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86691.260EA 32.97 EUR
86691.260 86691.180
VWR®, Manganese standard solution, 1000 mg/L Mn in dil. nitric acid AVS TITRINORM standard for AAS
Standaarden Kationische standaarden Mangaan standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%