VWR®, Selenium standard solution, 1,000 mg/L Se in dil. nitric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86709.260EA 23.1 EUR
86709.260 86709.180
VWR®, Selenium standard solution, 1,000 mg/L Se in dil. nitric acid AVS TITRINORM standard for AAS
Standaarden Kationische standaarden Selenium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%