Peha-taft® Latex Powderfree Gloves
Leverancier: HARTMANN PAUL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
942595 9425894 9425904 9425914 9425924 9425934 9425944 9425954 9425964
HART942595EA 98.9 EUR
HART942595 HART9425894 HART9425904 HART9425914 HART9425924 HART9425934 HART9425944 HART9425954 HART9425964
Peha-taft® Latex Powderfree Gloves
Handschoenen
Powder-free disposable surgical gloves made of thin and particularly soft latex, particularly easy to put on thanks to the special polymer inner coating, with a special micro-roughened surface for optimal grip, very good tactile sensitivity, highly elastic and tear-resistant, fatigue-free fit thanks to the fully anatomical fit.
  • Sophisticated
  • Refined and smooth without being slippery
  • Sterile
  • sealed in pairs with ozone protection
Peha-taft® latex is a medical product in accordance with EN 455 and personal protective equipment in accordance with EN 374, suitable for use with cytostatics and chemicals.
Suitable for all surgical procedures, including operations with a high risk of perforation (orthopedics, traumatology, accident surgery) and for high-risk patients as an overglove during double-gloving.
Recommended as an overglove in combination with Peha-underglove latex.
It achieves an AQL level of 0,65.
Verpakking: One folding box of 50, one box of 6 folding boxes.