Eszopiclone
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
467020010
467022500
467020010.EA
448
EUR
467020010.
467022500.
Eszopiclone
Eszopiclone
Formule:
C₁₇H₁₇ClN₆O₃ MW: 388.81 g/mol |
MDL Nummer:
MFCD03700720 CAS nummer: 138729-47-2 |
Specification Test Results
Appearance | white to light yellow powder |
Infrared spectrum | Conforms |
HPLC | 0.99 |
Loss on drying | ≤0.5 % (105°C, 3 hr) (vacuum) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...