Hexahydroindan
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
468530010 468532500
468530010.EA 452 EUR
468530010. 468532500.
Hexahydroindan
Hexahydroindan
Formule: C₉H₁₆
MDL Nummer: MFCD00021262
CAS nummer: 496-10-6
Order Now
Specification Test Results
Appearance clear colorless liquid
Infrared spectrum Conforms
GC ≥96.0 % (cis/trans)