2-Amino-6-methoxypurine 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

470270010 470270050 470270250
470270010.EA 62.9 EUR
470270010. 470270050. 470270250.
2-Amino-6-methoxypurine 95%
2-Amino-6-methoxypurine
Formule: C₆H₇N₅O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133009
CAS nummer: 20535-83-5

Order Now

Specification Test Results

Appearance white to light yellow to beige crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %
Water ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR