2-Iodonaphthalene 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
471040010
471040050
471040010.EA
97.5
EUR
471040010.
471040050.
2-Iodonaphthalene 98%
2-Iodonaphthalene
Formule:
C₁₀H₇I Kookpunt: 307,5°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00021590 CAS nummer: 612-55-5 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | white to yellow to brown crystalline powder and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 52°C to 56°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...