Potassium disulphate 95%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Dipotassium disulphate
472460025 472465000
472460025.EA 219 EUR
472460025. 472465000.
Potassium disulphate 95%, pure
Potassium disulphate
Formule: K₂O₇S₂
MW: 254.32 g/mol
Smeltpunt: 325
Dichtheid: 2.51
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011385
CAS nummer: 7790-62-7
EINECS: 232-216-8
UN: 2923
ADR: 8 I
Merck Index: 13,07749
Order Now
Specification Test Results
Appearance white adhering crystalline powder
Water ≤5.0 %
Titration with NaOH 94.0 to 106.0 %