Glucan (from yeast)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: beta-D-Glucan, Glucosan

472981000 472985000
472981000.EA 86.5 EUR
472981000. 472985000.
Glucan (from yeast)
Glucan
Formule: (C₆H₁₀O₅)n
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00466918
CAS nummer: 9012-72-0
EINECS: 232-739-1

Order Now

Specification Test Results

Appearance light yellow to brown powder or crystals
Infrared spectrum Conforms
Loss on drying ≤10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR