Dibenzyl toluene mixture of isomers 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Marlotherm SH
473440010 473442500
473440010.EA 83.8 EUR
473440010. 473442500.
Dibenzyl toluene mixture of isomers 97%
Dibenzyl toluene
Formule: C₂₁H₂₀
MW: 272.39 g/mol
MDL Nummer: MFCD01761976
CAS nummer: 53585-53-8
Order Now
Specification Test Results
Appearance colorless liquid
Infrared spectrum Conforms
HPLC ≥96 % (total isomers)