3,4'-Diaminodiphenyl ether 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 3-(4-Aminophenoxy)aniline, 3,4'-Oxydianiline
474460010 474460050 474460250
474460010.EA 33.2 EUR
474460010. 474460050. 474460250.
3,4'-Diaminodiphenyl ether 98%
3,4'-Diaminodiphenyl ether
Formule: C₁₂H₁₂N₂O
Kookpunt: 385°C (760mmHg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036097
CAS nummer: 2657-87-6
EINECS: 220-190-0
Order Now
Specification Test Results
Appearance white to brown powder or crystals
Infrared spectrum Conforms
GC ≥97.5 %
Titration with HClO4 97.5 to 102.5 %