Insyte™ Autoguard™ IV Catheters
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
381237 381334 381212 381257 381223 381267 381934 381244 381247 381323 381312 381837 381234 381823 381847
BDAM381237EA 521 EUR
BDAM381237 BDAM381334 BDAM381212 BDAM381257 BDAM381223 BDAM381267 BDAM381934 BDAM381244 BDAM381247 BDAM381323 BDAM381312 BDAM381837 BDAM381234 BDAM381823 BDAM381847
Insyte™ Autoguard™ IV Catheters
Katheters
The BD Insyte™ Autoguard™ Shielded IV catheter incorporates advanced safety features, including a push-button that instantly retracts the needle, helping reduce the risk of accidental needlestick injuries.
  • BD push-button needle shielding technology is demonstrated to reduce needlestick injuries by 95%
  • BD Instaflash™ needle technology on 20 - 24 G products may improve first-stick success and reduce painful hit-and-miss insertions
  • BD Vialon™ biomaterial softens up to 70% in the vein, enabling longer dwell times and reducing the chance of mechanical phlebitis
More Product Information
Order Now
Specificaties
Environmentally Preferable