Monsterbuisjes
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
114715 114760 114750
212-1744EA 112 EUR
212-1744 212-0294 212-0441
Monsterbuisjes
Buisjes Reageerbuisjes
PS, crystal clear.
  • Robust
Universal tubes for a wide range of applications.
Bestelinformatie: Fitting stoppers must be ordered separately.
Order Now
Specificaties
Environmentally Preferable