Corning® Synthegel™ Synthetic Hydrogel Matrix Kits
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
354787 354789 354791 354792
BDAA354787EA 403 EUR
BDAA354787 BDAA354789 BDAA354791 BDAA354792
Corning® Synthegel™ Synthetic Hydrogel Matrix Kits
Celcultuurmedia
Corning® Synthegel™ 3D matrices are chemically defined peptides that self assemble into nanofibers which crosslink to form tuneable hydrogels. These hydrogels provide a robust and reproducible microenvironment for 3D culture of physiological spheroids and hiPS cells. The Synthegel™ matrix kits provide negligible variability and a more controlled substrate for in vivo-like 3D cell culture. Such a platform is needed for cancer research, stem cell research, and drug screening in a 3D format.
  • Matrix rigidity is tunable by altering the peptide concentration
  • Purified synthetic peptide in neutral ph
  • Cells are encapsulated in physiological mimicking conditions
  • Fast hydrogel formation (5 to 30 minutes)
  • Sol-gel transfer reversible by shearing force
  • Works with standard cell culture methodologies, growth factors and media
  • Allows growth and passaging of hiPSCs in 3D format. The hiPSC Grow mix supplement provides a complete environment for hiPSC 3D culture
  • Allows 3D embedded and suspension hiPSC cultures
Corning® Synthegel™ 3D matrix kits are chemically defined, self-healing synthetic hydrogel bio-tools for culturing many different types of cells, including cancer and stem cells, in a 3D format with robustness and convenience. The Synthegel™ 3D matrix platform supports culture of hiPSC in a 3D embedded format or in an encapsulation for suspension format. It also supports the culture of high content physiological cancer spheroids within a defined, tuneable, and synthetic peptide matrix devoid of acidic or chilled conditions. The 3D matrix peptide solution is designed to create an extracellular matrix mimicking hydrogel in one simple mixing step. The Synthegel™ 3D matrix solution is formulated in standard cell culture media at neutral pH and is stable at room temperature. The hydrogel formation can be initiated by directly mixing the peptide solution with the cross-linking agent along with any cell suspension without extra steps. Cultured cells are easy to harvest. The Synthegel™ 3D matrix kits provide excellent reproducibility and work with standard biological methodologies.
Certificaten: Complies with ISO 9001. This product is classified for use as Research Use Only (RUO).
Bestelinformatie: The Synthegel™ Spheroid matrix kit contains Synthegel™ spheroid matrix and X-linking solution. The Synthegel™ 3D hiPSC matrix kit contains Synthegel™ 3D hiPSC matrix and X-linking solution. The Synthegel™ 3D hiPSC Suspension Matrix Kit contains Synthegel™ 3D hiPSC Suspension matrix and X-linking solution.
The Synthegel™ 3D hiPSC matrix kit and the Synthegel™ 3D hiPSC Suspension matrix kit requires a vial of lyophilised Synthegel™ 3D hiPSC Grow Mix, 1 mg.
Leveringsinformatie: All matrix kits are shipped at ambient temperature and should be refrigerated upon arrival. The grow mix is shipped in dry ice and should be frozen upon arrival.