QUICKFIT® Weighing Bottles, Clear, DWK Life Sciences
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WB25/40 WB30/50 WB30/60 WB35/30 WB35/70 WB40/80 WB50/30 WB50/80
BIBBWB25/40EA 22.3 EUR
BIBBWB25/40 BIBBWB30/50 BIBBWB30/60 BIBBWB35/30 BIBBWB35/70 BIBBWB40/80 BIBBWB50/30 611-2566
QUICKFIT® Weighing Bottles, Clear, DWK Life Sciences
Flessen Weegflessen
Manufactured from borosilicate glass 3.3, these weighing bottles are supplied complete with ground lid.
  • Lightweight construction with ground glass stoppers
  • Choice of squat or tall form weighing bottles
  • Supplied complete with ground lid
  • WB35/30 and WB50/80 are supplementary to DIN range
In order to carry out reactions in a controlled manner, it is essential to know the quantities of substances required and to weigh the corresponding substances accurately. A weighing glass can be used for weighing. The associated lid allows the weighing pan to be closed and thus prevents, for example, turbulence of the weighed-in solids during transport.
Since even the smallest contamination of the weighing glass can have an effect on the weighing inheritance, the glass in the area of the ground joint and the lid knob are so pronounced that they can be grasped and moved with forceps or tweezers.
Certificaten:
Complies with DIN 12605.