IPTG (isopropyl-β-D-thiogalactopyranoside)
Leverancier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: IPTG
05684-1 05684-2 05684-3
BMOL05684-1EA 0 EUR
BMOL05684-1 BMOL05684-2 BMOL05684-3
IPTG (isopropyl-β-D-thiogalactopyranoside)
IPTG (isopropyl-β-D-thiogalactopyranoside)
Formule: C₉H₁₈O₅S
MW: 238.31 g/mol
Kookpunt: 402
Smeltpunt: 109.5…110.5 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00063273
CAS nummer: 367-93-1
EINECS: 206-703-0