Nuclear Fast Red 94%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
211980010 211980050
ACRO211980010EA 37.1 EUR
ACRO211980010 ACRO211980050
Nuclear Fast Red 94%, pure
Nuclear Fast Red
Formule: C₁₄H₈NNaO₇S
MW: 357.28 g/mol
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00078493
CAS nummer: 6409-77-4
EINECS: 229-088-0
Merck Index: 13,06512
Order Now
Specification Test Results
Appearance Dark red to bordeaux Powder
Infrared spectrum Conforms
HPLC ≥92 %