Sudan III ≥90.0% (by UV/Vis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

S0142-25G S0142-100G
TCIAS0142-25GEA 22 EUR
TCIAS0142-25G TCIAS0142-100G
Sudan III ≥90.0% (by UV/Vis)
Sudan III
Formule: C₂₂H₁₆N₄O
MDL Nummer: MFCD00003905
CAS nummer: 85-86-9
EINECS: 201-638-4
Merck Index: 14,08884
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=652a7f66bb70072b318c95c7c4a71c94cfc7b26b&vid=652a7f66bb70072b318c95c7c4a71c94cfc7b26b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (UV-vis method) min. 90.0 %
Lambda max. 511.0 to 515.0 nm (CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR