Pyridoxine hydrochloride
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI
33990.01 33990.02 33990-0025
SERA33990.01EA 0 EUR
SERA33990.01 SERA33990.02 SERA33990-0025
Pyridoxine hydrochloride
Pyridoxine hydrochloride
Formule: C₈H₁₂ClNO₃
Smeltpunt: 214…215 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012807
CAS nummer: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072