Lanthanum(III) nitrate

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

27507.04 27507.01
SERA27507.04EA 0 EUR
SERA27507.04 SERA27507.01
Lanthanum(III) nitrate
Lanthanum(III) nitrate
Formule: LaN₃O₉
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011067
CAS nummer: 10099-59-9
EINECS: 233-238-0
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR