Methyl orange - Bromocresol green in aqueous solution Reag. Ph. Eur. 1054801 mixed indicator
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87870.180EA 54.3 EUR
87870.180
Methyl orange - Bromocresol green in aqueous solution Reag. Ph. Eur. 1054801 mixed indicator
Methyl orange - Bromocresol green
Colour change: pH 3.0 (orange) to pH 4.4 (olive-green)
The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for Purified Water.
Bestelinformatie: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022
Order Now
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022