CHIRALPAK® IM HPLC Columns
Leverancier: CHIRAL TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
92G22 92G24 92G25 92G35 92G45 92G94
DAIC92G22EA 638 EUR
DAIC92G22 DAIC92G24 DAIC92G25 DAIC92G35 DAIC92G45 DAIC92G94
CHIRALPAK® IM HPLC Columns
Kolommen voor chromatografie
The CHIRALPAK® IM-10, 10 µm guard column is compatible with SFC mode (tested), HPLC (Normal and Reversed-Phase), and supports analytical techniques using guards, cartridges, and holders.