Flat Cut Rimmed Bowls, 304 Grade Stainless Steel
Leverancier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOW4003P BOW4004P
COLDBOW4003PEA 77.4 EUR
COLDBOW4003P COLDBOW4004P
Flat Cut Rimmed Bowls, 304 Grade Stainless Steel
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Stainless steel bowls available in 304 grade material, ideal for laboratory and other pharmaceutical industry uses.
  • Hygienic design; pressed in one piece, with a flat base and flat rim
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Optional material grade, surface finish, and TSE certification available upon request
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information.