Uvex Silv-Air Premium Respirators, FFP 2
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8735321 8735311 8735211 8735310 8735101 8735111 8735200 8735221 8735210 8735110 8735201 8735100
UVEX8735321EA 122 EUR
UVEX8735321 UVEX8735311 UVEX8735211 UVEX8735310 UVEX8735101 UVEX8735111 UVEX8735200 UVEX8735221 UVEX8735210 UVEX8735110 UVEX8735201 UVEX8735100
Uvex Silv-Air Premium Respirators, FFP 2
Ademhalingsmaskers Wegwerpmaskers Stofmaskers
The Uvex silv-Air premium product line comprises classic, white folding respirators that adapt to different face shapes. The headband and integrated sealing lip in all protection classes mean the masks are easy to put on correctly as well as making them comfortable to wear, even for extended periods.
  • Particle-filtering folding mask for small to medium and larger face shapes
  • White colour
  • Compatible with Uvex safety eyewear
  • Adjustable head band offering easy handling and comfortable fit
  • Wide, seamless headband for ensuring a comfortable and secure fit
  • Incorporated nose clip that is easy to adjust
  • Comfortable sealing lip in the nose area for a reliable fit
  • Exhalation valve for reducing heat and humidity as well as noticeably reducing breathing resistance
FFP2 respirators are suitable for smoke, particles and dusts that are noxious or harmful to health. They filter at least 94% of particles and can be used up to ten times the respective occupational exposure limit.
With a filtering efficiency of at least 99%, FFP3 masks constitute the highest protection class. They filter up to 30 times the respective occupational exposure limit and are suitable for protecting against toxic particles, dusts and smoke.
5210+/ 5220+: exhalation valve for reducing heat and humidity as well as noticeably reducing breathing resistance
5220+: additional activated carbon filter for protection against odours, gases and vapours under the respective limit values
Certificaten: EN 149:2001 and A1:2009.
Verpakking: Individually packed.