Formalin 10% Ready-to-Use, buffered (Neutral)

Leverancier: BIO OPTICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Formaldehyde

05-01V3000P 05-01V5000P 05-01V500P
BOPT05-01V3000PEA 137 EUR
BOPT05-01V3000P BOPT05-01V5000P BOPT05-01V500P
Formalin 10% Ready-to-Use, buffered (Neutral)
Formalin
Formule: CH₂O
MW: 30.03 g/mol
Kookpunt: 100 °C
Vlampunt: >60 °C
Bewaar temperatuur: Room Temperature
MDL Nummer: MFCD00003274
CAS nummer: 50-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR