DURAN® GLS 80® Production and Storage Bottles
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
219918685 219919184
SCOT219918685EA 948 EUR
SCOT219918685 SCOT219919184
DURAN® GLS 80® Production and Storage Bottles
Flessen Glazen flessen
Graduated, manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements.
  • Thick-walled glass and therefore mechanically more resistant
  • The external PU polymer coating provides scratch protection
  • The flat, robust base allows the use of large and heavy stir bars
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • Optimum filling and particularly easy cleaning
Bestelinformatie: Supplied as safety coated glass bottle only with a HDPE push-on dust cover. The screw caps and pouring rings must be ordered separately.