Light Duty Measuring Jugs
Leverancier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MEA4001G MEA4002G MEA40.5G
COLDMEA4001GEA 51.9 EUR
COLDMEA4001G COLDMEA4002G COLDMEA40.5G
Light Duty Measuring Jugs
Bekers
Stainless steel measuring jugs meet international pharma quality compliance regulations.
  • All sizes available as standard, off-the-shelf for delivery within days
  • 304 pharmaceutical grade stainless steel
  • Graduated in pints and litres
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Material Grade, Surface Finish, and TSE certification available upon request
Made from 304 grade stainless steel, these measuring jugs are tapered and come with either an open or closed handle. They feature crevice-free and pit-free interiors and are bright polished throughout.
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information. Measuring jugs are available in Heavy Duty, Light Duty, and Light Duty with rimmed foot formats. The option of rimmed feet is available with the closed handle version from 0.5 litre upwards.