Stainless Steel Cans with Lid
Leverancier: Adelphi Pharma Hygiene Products
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CWL4010P CWL4001P
COLDCWL4010PEA 577 EUR
COLDCWL4010P COLDCWL4001P
Stainless Steel Cans with Lid
Monsternamepotjes Containers voor algemeen gebruik
A range of hygienic, high-quality stainless steel cans which meet international pharma and food quality compliance regulations.
  • Choose from 304 or 316L pharmaceutical grade stainless steel
  • Containers available off-the-shelf for delivery within days
  • Exceptional average surface finish of 0.4 mm Ra
  • Plain or graduated (graduations are available for sizes 5 L and above)
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Optional Material Grade, Surface Finish, and TSE certification available upon request
These cans come complete with lids and swing handles. They feature crevice-free and pit-free interiors and are bright polished throughout.
A range of optional modifications are available to meet every requirement, including airtight toggle clamps and seals, mobile frames for decanting, bespoke etching of logos and other information, and more.