BioTherm Dry Ice Systems (Category B Compliant)
Leverancier: DGP INTELSIUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BT104 BT111
DGINBT104EA 63.7 EUR
DGINBT104 DGINBT111
BioTherm Dry Ice Systems (Category B Compliant)
Mailer enveloppen en transportverpakkingen
The BioTherm Dry Ice range is designed forthe safe transportation of deep frozen samples to IATA, ADR and DOT (49 CFR) transportregulations. The systems offer a variety of options for the safe transit of Category B shipments, as complete systems or as overpacks.
  • Infectious samples
  • Diagnostic samples
  • Clinical trials
  • Screening programmes
  • Eradication schemes
Category B systems provided with compliant transport bags and absorbent.