Chemicon® ReBlot Plus Kits
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2500-S 2500
MILF2500-SEA 120 EUR
MILF2500-S MILF2500
Chemicon® ReBlot Plus Kits
Reagentia voor elektroforese Western Blotting Reagents
Western blotting is a commonly used technique for studying protein function and localisation. Typically, protein samples are electrophoresed on SDS-page and transferred to a membrane such as nitrocellulose or nylon, where they are probed with specific antibodies. Unlike nucleic acid based technologies, which allow reuse of southern and northern blots, it has been difficult to reuse western blots.
  • Detection method: Chemiluminescence
  • Mild antibody stripping solution (10X) - (1 container, 50 ml)
  • Strong antibody stripping solution (10X) - (1 container, 50 ml)
The MILLIPORE re-blot plus western blot recycling kit is effective for removal of antibodies from western blots that have been developed with chemiluminescence or radioactive iodine or other isotopes. It is not recommended for stripping colorimetric substrates (TMB, DAB, 4-chloronapthol, and more.), as it is not possible to effectively remove substrates that precipitate at the reaction site.
The re-blot western blot recycling kit should be used only for qualitative purposes until it has been established by comparative blot analysis that stripping does not quantitatively affect a given antigen.
Opgelet: Note: To prevent reagent degradation secure the cap tightly upon storage. Avoid extended exposure to air.