Millicell® 24 Wells Cell Culture Insert Plates
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PSRP010R1 PSHT010R5 PSMT010R1 PSST010R5 PSET010R5 PSRP010R5 PSHT010R1 PSET010R1 PSST010R1 PSMT010R5
MILFPSRP010R1EA 176 EUR
MILFPSRP010R1 MILFPSHT010R5 MILFPSMT010R1 MILFPSST010R5 MILFPSET010R5 MILFPSRP010R5 MILFPSHT010R1 MILFPSET010R1 MILFPSST010R1 MILFPSMT010R5
Millicell® 24 Wells Cell Culture Insert Plates
Microplaten Celcultuurplaten
Millicell cell culture insert plates are a patented device designed to support cell growth and differentiation of endothelial and epithelial cell lines, including CaCo-2.
  • Isopore™ matrix
  • 0,7 cm² filtration area
  • 9,5 mm filter diameter
  • Tissue Culture (TC)-treated surface binding type
  • Dimension: 84,8×128×26,2 mm
Seed-and-feed systems
Automated liquid handling systems and span pipettes
Basolateral and apical access to cells
Transepithelial electrical resistance (TEER) measurement systems
The Millicell device is available with a 0,4 µm PCF and a variety ofMore Product Information