MultiScreenHTS Filter Plates with Glass Fibre Filter
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MZFBN0W50 MSFBN6B10 MSFBN6B50 MZFBN0W10 MSFCN6B50 MSFCN6B10 MZFCN0W10 MSFBNXB50 MZFCN0W50
MILFMZFBN0W50EA 3690 EUR
MILFMZFBN0W50 MILFMSFBN6B10 MILFMSFBN6B50 MILFMZFBN0W10 MILFMSFCN6B50 MILFMSFCN6B10 MILFMZFCN0W10 MILFMSFBNXB50 MILFMZFCN0W50
MultiScreenHTS Filter Plates with Glass Fibre Filter
Microplaten Filtreerplaten
MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96 and 384-well platforms with a broad selection of membranes and plastics.
  • Rigid sidewalls improve handling
  • Ample surfaces are provided for bar code labels
MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.
The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.