MultiScreen Solvinert Filter Plates with PTFE Membrane, 96-well
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MSRLN0450 MSRPN0410 MSRPN0450 MSRLN0410 MDRLN0410 MDRPNP410
MILFMSRLN0450EA 1780 EUR
MILFMSRLN0450 MILFMSRPN0410 MILFMSRPN0450 MILFMSRLN0410 MILFMDRLN0410 MILFMDRPNP410
MultiScreen Solvinert Filter Plates with PTFE Membrane, 96-well
Microplaten Filtreerplaten
MultiScreen HTS 96-well filtration plates are used to filter samples and perform entire procedures, from cell growth to scintillation counting within the same plate.
  • PTFE membrane (hydrophilic or hydrophobic)
  • Flat bottom wells
  • Non sterile
  • Polyolefin copolymer
The plates come in many membrane types, pore sizes, and plate materials. The automation-compatible design allows for easy manipulation with a variety of robotics systems and makes barcoding possible on all four sides. Filtration is accomplished either by vacuum using the MultiScreen HTS VacuumMore Product Information