Millex® Syringe Filters, LCR Hydrophilic PTFE
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SLCR033NS SLCR033NB SLCR033NK SLLG033NS SLLG033NB SLLG033NK SLCRX13NK SLCRX13NL SLCRX13TL SLLGX13NK SLLGX13NL
MILFSLCR033NSEA 333 EUR
MILFSLCR033NS MILFSLCR033NB MILFSLCR033NK MILFSLLG033NS MILFSLLG033NB MILFSLLG033NK MILFSLCRX13NK MILFSLCRX13NL MILFSLCRX13TL MILFSLLGX13NK MILFSLLGX13NL
Millex® Syringe Filters, LCR Hydrophilic PTFE
Filters Spuitfilters
Millex® Syringe filters are HPLC-certified for low levels of UV-absorbing extractables.
  • Lowest extractables, optimizing background levels of sensitive analyses like UHPLC and LC-MS
  • Filtration of protein-containing aqueous or organic solutions
  • Available in 0,20 μm and 0,45 μm pore sizes and three diameters to suit your application needs
  • Polypropylene housing material offers low levels of extractables and broad chemical compatibility
Applications are sample filtration prior to UHPLC, HPLC, LC-MS, and mass spec, Solvent filtration, Clarification ofMore Product Information